Anti-CD183 (CXCR3) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: CXCR3-173]

Supplier: BioLegend
126523 126523
10759-290EA 271.58 USD
CA10759-290 10759-290
Anti-CD183 (CXCR3) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: CXCR3-173]
Antibodies
Brilliant Violet 605™ anti-mouse CD183 (CXCR3) [CXCR3-173]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD183 (CXCR3)
Clonality: Monoclonal
Clone: CXCR3-173
Conjugation: Brilliant Violet® 605
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR