Anti-CD183 Mouse Monoclonal Antibody (LEAF) [clone: G025H7]

Supplier: BioLegend
353717 353718 353718 353717
353718-BLEA 425.33 USD
CA353717-BL CA353718-BL 353718-BL 353717-BL
Anti-CD183 Mouse Monoclonal Antibody (LEAF) [clone: G025H7]
LEAF™ Purified anti-human CD183 (CXCR3) [G025H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: Block, FC; Size: 500 μg

Each lot of this antibody is quality control tested by blocking. Chemotaxis of human CXCR3-transfected BaF3 cells was performed in the presence of 20 ng/ml of recombinant CXCL11. Addition of LEAF™ purified anti-human CXCR3 (clone G025H7) inhibits migration of hCXCR3 transfected BaF3 cells (ED50=0.15 μg/ml).

Type: Primary
Antigen: CD183
Clonality: Monoclonal
Clone: G025H7
Conjugation: LEAF
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR