Anti-CD18 Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: M18/2]

Supplier: BioLegend
101416 101416
10756-752EA 273.08 USD
CA10756-752 10756-752
Anti-CD18 Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: M18/2]
Antibodies
Alexa Fluor® 594 anti-mouse CD18 [M18/2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunohistochemisty, a concentration range of 2-5 μg/ml is suggested. For immunofluorescence microscopy, a concentration range of 2-10 μg/ml is recommended. <span>For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume.</span> It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: M18/2
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR