Anti-CD172a Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: P84]

Supplier: BioLegend
144012 144012 144011 144011
10757-348EA 139.18 USD
CA10757-348 10757-348 CA10757-350 10757-350
Anti-CD172a Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: P84]
Antibodies
PE anti-mouse CD172a (SIRPα) [P84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD172a
Clonality: Monoclonal
Clone: P84
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR