Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD172a Rat Monoclonal Antibody (LEAF) [clone: P84]

Supplier: Biolegend

144004 144003 144004 144003
CA144004-BL 144003-BL 144004-BL CA144003-BL
Anti-CD172a Rat Monoclonal Antibody (LEAF) [clone: P84]
Antibodies
LEAF™ Purified anti-mouse CD172a (SIRPα) [P84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC, Block, IF, IHC, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD172a
Clonality: Monoclonal
Clone: P84
Conjugation: LEAF
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty