Anti-CD169 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: 3D6.112]

Supplier: BioLegend
142413 142413
10757-284EA 327.4 USD
CA10757-284 10757-284
Anti-CD169 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: 3D6.112]
Antibodies
Brilliant Violet 605™ anti-mouse CD169 (Siglec-1) [3D6.112]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD169
Clonality: Monoclonal
Clone: 3D6.112
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR