Anti-CD161 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3.2.3]

Supplier: BioLegend
205606 205606
205606-BLEA 298.35 USD
CA205606-BL 205606-BL
Anti-CD161 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3.2.3]
Antibodies
APC anti-rat CD161 [3.2.3]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: 3.2.3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR