Anti-CD160 Rat Monoclonal Antibody [clone: 7H1]

Supplier: BioLegend
143002 143002
143002-BLEA 191.54 USD
143002-BL CA143002-BL
Anti-CD160 Rat Monoclonal Antibody [clone: 7H1]
Antibodies
Purified anti-mouse CD160 [7H1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD160
Clonality: Monoclonal
Clone: 7H1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR