Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD16 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: B73.1]

Supplier: Biolegend

360713 360714 360713 360714
CA10760-412 10760-414 10760-412 CA10760-414
Anti-CD16 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: B73.1]
Antibodies
Alexa Fluor® 647 anti-human CD16 [B73.1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: B73.1
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty