Anti-CD16 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: B73.1]

Supplier: BioLegend
360709 360710 360710 360709
10760-406EA 390.48 USD
CA10760-404 10760-406 CA10760-406 10760-404
Anti-CD16 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: B73.1]
Antibodies
APC/Cy7 anti-human CD16 [B73.1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: B73.1
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR