Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSNK1A1 Rabbit Polyclonal Antibody

Supplier: Abgent

AP14783B
89520-986EA 418.66 USD InStock
89520-986
Anti-CSNK1A1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Flow Cytometry: 1:10~50,

Type: Primary
Antigen: CSNK1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty