Anti-CD16 Mouse Monoclonal Antibody [clone: B73.1]

Supplier: BioLegend
360702 360701 360701 360702
10760-430EA 133.83 USD
CA10760-430 10760-430 10760-748 CA10760-748
Anti-CD16 Mouse Monoclonal Antibody [clone: B73.1]
Antibodies
Purified anti-human CD16 [B73.1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: B73.1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR