Anti-TXNIP Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20130C
89513-860EA 636.79 USD
89513-860
Anti-TXNIP Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: TXNIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR