Anti-CDC20B Rabbit Polyclonal Antibody

Supplier: Abcepta
AP17515A
89518-236EA 636.61 USD
89518-236
Anti-CDC20B Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CDC20B
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR