Anti-CD155 (PVR) Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: TX56]

Supplier: BioLegend
131511 131511 131512 131512
10757-170EA 422.45 USD
10757-170 10757-168 CA10757-168 CA10757-170
Anti-CD155 (PVR) Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: TX56]
Antibodies
PE/Cy7 anti-mouse CD155 (PVR) [TX56]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD155 (PVR)
Clonality: Monoclonal
Clone: TX56
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR