Anti-FBXO11 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP16914A
89518-832EA 606.46 USD
89518-832
Anti-FBXO11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: FBXO11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR