Anti-CD152 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UC10-4B9]

Supplier: BioLegend
106314 106314 106313 106313
10756-876EA 416.98 USD
10756-876 10756-874 CA10756-874 CA10756-876
Anti-CD152 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UC10-4B9]
Antibodies
PE/Cy7 anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD152
Clonality: Monoclonal
Clone: UC10-4B9
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR