Anti-CD152 Mouse Monoclonal Antibody (LEAF) [clone: L3D10]

Supplier: BioLegend
349903 349903 349904
349903-BLEA 152.88 USD
349903-BL CA349903-BL CA349904-BL
Anti-CD152 Mouse Monoclonal Antibody (LEAF) [clone: L3D10]
LEAF™ Purified anti-human CD152 (CTLA-4) [L3D10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, Block; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD152
Clonality: Monoclonal
Clone: L3D10
Conjugation: LEAF
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR