Anti-CD151 Mouse Monoclonal Antibody (LEAF) [clone: 50-6]

350404 350404
350404-BLEA 331.97 USD
350404-BL CA350404-BL
Anti-CD151 Mouse Monoclonal Antibody (LEAF) [clone: 50-6]
LEAF™ Purified anti-human CD151 (PETA-3) [50-6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, Block, ICC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: 50-6
Conjugation: LEAF
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR