Anti-CD150 (SLAM) Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: TC15-12F12.2]

Supplier: BioLegend
115929 115929
10756-934EA 243.76 USD
CA10756-934 10756-934
Anti-CD150 (SLAM) Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: TC15-12F12.2]
Antibodies
Brilliant Violet 510™ anti-mouse CD150 (SLAM) [TC15-12F12.2]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD150 (SLAM)
Clonality: Monoclonal
Clone: TC15-12F12.2
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: Rat IgG2a, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR