Anti-CD15 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: W6D3]

Supplier: BioLegend
323037 323038 323037 323038
10758-278EA 456.05 USD
10758-278 CA10758-276 CA10758-278 10758-276
Anti-CD15 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: W6D3]
Antibodies
PE/Dazzle™ 594 anti-human CD15 (SSEA-1) [W6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: W6D3
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR