Anti-TMEM200A Rabbit Polyclonal Antibody

Supplier: Abcepta
AP16298A
89520-172EA 636.79 USD
89520-172
Anti-TMEM200A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: TMEM200A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR