Anti-CD146 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ME-9F1]

Supplier: BioLegend
134713 134714 134714 134713
10756-386EA 414.63 USD
CA10756-386 CA10756-384 10756-386 10756-384
Anti-CD146 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ME-9F1]
Antibodies
PE/Cy7 anti-mouse CD146 [ME-9F1]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: ME-9F1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR