Anti-CD146 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1H12]

Supplier: BioLegend
361005 361006 361005 361006
10760-806EA 289.19 USD
10760-806 10760-804 CA10760-804 CA10760-806
Anti-CD146 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1H12]
Antibodies
PE anti-human CD146 [P1H12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: P1H12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR