Anti-CD146 Mouse Monoclonal Antibody [clone: P1H12]

Supplier: BioLegend
361002 361001 361002 361001
10760-800EA 135.97 USD
CA10760-798 CA10760-800 10760-800 10760-798
Anti-CD146 Mouse Monoclonal Antibody [clone: P1H12]
Antibodies
Purified anti-human CD146 [P1H12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: P1H12
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR