Anti-CD34 Mouse Monoclonal Antibody [clone: B-G25]

Supplier: Cell Sciences
CDM176
102995-400EA 443.08 USD
102995-400
Anti-CD34 Mouse Monoclonal Antibody [clone: B-G25]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: B-G25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR