Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD146 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: ME-9F1]

Supplier: Biolegend

134708 134707 134708 134707
134708-BLEA 280.83 USD InStock
134708-BL 134707-BL CA134708-BL CA134707-BL
Anti-CD146 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: ME-9F1]
Antibodies
Alexa Fluor® 488 anti-mouse CD146 [ME-9F1]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunohistochemistry, a concentration range of 5.0 - 10.0 μg/ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: ME-9F1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty