Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD144 Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: BV9]

Supplier: Biolegend

348512 348512
CA10759-382 10759-382
Anti-CD144 Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: BV9]
Antibodies
Alexa Fluor® 594 anti-human CD144 (VE-Cadherin) [BV9]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: IF, IHC-P; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 5-10 μg/ml is recommended. <span>For immunohistochemistry, a concentration range of 5 - 10 μg/ml is suggested. </span>It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD144
Clonality: Monoclonal
Clone: BV9
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty