Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: HCD14]

Supplier: BioLegend
325633 325633 325634 325634
10759-492EA 152.22 USD
CA10759-494 CA10759-492 10759-492 10759-494
Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: HCD14]
Antibodies
PE/Dazzle™ 594 anti-human CD14 [HCD14]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: HCD14
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR