Anti-RALYL Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20112A
89514-462EA 637.11 USD
89514-462
Anti-RALYL Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: RALYL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR