Anti-LGALS9 Mouse Monoclonal Antibody [clone: B-K38]

Supplier: Cell Sciences
CDM453
102995-442EA 553.85 USD
102995-442
Anti-LGALS9 Mouse Monoclonal Antibody [clone: B-K38]
Antibodies
Type: Primary
Antigen: LGALS9
Clonality: Monoclonal
Clone: B-K38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR