Anti-CD56 Mouse Monoclonal Antibody [clone: B-A19]

Supplier: Cell Sciences
CDM047
102995-506EA 553.85 USD
102995-506
Anti-CD56 Mouse Monoclonal Antibody [clone: B-A19]
Antibodies
Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: B-A19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR