Anti-CD134 (OX-40) Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: OX-86]

Supplier: BioLegend
119411 119411
119411-BLEA 220.33 USD
CA119411-BL 119411-BL
Anti-CD134 (OX-40) Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: OX-86]
Antibodies
Brilliant Violet 421™ anti-mouse CD134 (OX-40) [OX-86]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD134 (OX-40)
Clonality: Monoclonal
Clone: OX-86
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR