Anti-CD134 Mouse Monoclonal Antibody [clone: Ber-ACT35 (ACT35)]

Supplier: BioLegend
350002 350002
350002-BLEA 127.42 USD
CA350002-BL 350002-BL
Anti-CD134 Mouse Monoclonal Antibody [clone: Ber-ACT35 (ACT35)]
Antibodies
Purified anti-human CD134 (OX40) [Ber-ACT35 (ACT35)]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, ELISA, IHC, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD134
Clonality: Monoclonal
Clone: Ber-ACT35 (ACT35)
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR