Anti-CD133 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 315-2C11]

Supplier: BioLegend
141210 141209 141210 141209
10757-238EA 427.87 USD
CA10757-238 10757-238 CA10757-236 10757-236
Anti-CD133 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 315-2C11]
Antibodies
PE/Cy7 anti-mouse CD133 [315-2C11]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD133
Clonality: Monoclonal
Clone: 315-2C11
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR