Anti-IL2RA Mouse Monoclonal Antibody [clone: B-B10]

Supplier: Cell Sciences
CDM001
102995-320EA 540 USD
102995-320
Anti-IL2RA Mouse Monoclonal Antibody [clone: B-B10]
Antibodies
Inhibits Mixed Lymphocyte Culture.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: B-B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR