Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD13 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: WM15]

Supplier: Biolegend

301715 301716 301716 301715
CA10757-662 10757-664 CA10757-664 10757-662
Anti-CD13 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: WM15]
Antibodies
Brilliant Violet 421™ anti-human CD13 [WM15]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cotton-topped Tamarin; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: WM15
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon;Chimpanzee;Cotton-topped Tamarin

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty