Anti-ICAM1 Mouse Monoclonal Antibody [clone: B-H17]

Supplier: Cell Sciences
CDM045
102995-340EA 553.85 USD
102995-340
Anti-ICAM1 Mouse Monoclonal Antibody [clone: B-H17]
Antibodies
Blocks PHA stimulated T cell proliferation. Does not block cell adhesion.

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: B-H17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR