Anti-CD120b Rat Monoclonal Antibody [clone: 3G7A02]

Supplier: BioLegend
358408 358408
10760-366EA 427.61 USD
CA10760-366 10760-366
Anti-CD120b Rat Monoclonal Antibody [clone: 3G7A02]
Antibodies
Ultra-LEAF™ Purified anti-human CD120b [3G7A02]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: Neut, FC; Size: 500 μg

Each lot of this antibody is quality control tested by neutralization. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD120b
Clonality: Monoclonal
Clone: 3G7A02
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR