Anti-CD120b Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3G7A02]

Supplier: BioLegend
358406 358405 358405 358406
10760-364EA 388.67 USD
CA10760-364 10760-364 10760-362 CA10760-362
Anti-CD120b Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3G7A02]
Antibodies
APC anti-human CD120b [3G7A02]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD120b
Clonality: Monoclonal
Clone: 3G7A02
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR