Anti-CD31 Mouse Monoclonal Antibody [clone: B-B38]

Supplier: Cell Sciences
CDM035
102995-490EA 553.85 USD
102995-490
Anti-CD31 Mouse Monoclonal Antibody [clone: B-B38]
Antibodies
Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: B-B38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR