Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD11c Armenian Hamster Monoclonal Antibody [clone: N418]

Supplier: Biolegend

117341 117341
10758-876EA 103.33 USD InStock
10758-876 CA10758-876
Anti-CD11c Armenian Hamster Monoclonal Antibody [clone: N418]
Antibodies
Purified anti-mouse CD11c (MaxPar® Ready) [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, CyTOF®; Size: 100 μg

This product is suitable for use with the <a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the <a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty