Anti-EPM2AIP1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20528A
10217-798EA 580.86 USD
10217-798
Anti-EPM2AIP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: EPM2AIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR