Anti-CD8A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-Z31]

Supplier: Cell Sciences
CDM252
102995-418EA 609.23 USD
102995-418
Anti-CD8A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-Z31]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: B-Z31
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR