Anti-CD11b Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: M1/70]

Supplier: BioLegend
101254 101254
10756-738EA 332.64 USD
CA10756-738 10756-738
Anti-CD11b Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: M1/70]
Antibodies
Alexa Fluor® 594 anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit (Lapine); Apps: IHC, IF, FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunohistochemisty, a concentration range of 2-5 μg/ml is suggested. For immunofluorescence microscopy, a concentration range of 2-10 μg/ml is recommended. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cynomolgus;Rhesus;Baboon;Chimpanzee,Rabbit (Lapine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR