Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD11b Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: M1/70]

Supplier: Biolegend

101239 101239
101239-BLEA 230.51 USD InStock
101239-BL CA101239-BL
Anti-CD11b Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: M1/70]
Antibodies
Brilliant Violet 650™ anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit (Lapine); Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is  0.3 μg per million cells in 100 μl volume. For flow cytometric staining using the test size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cynomolgus;Rhesus;Baboon;Chimpanzee,Rabbit (Lapine)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty