Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD11b Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: ICRF44]

Supplier: Biolegend

301340 301340
10757-614 CA10757-614
Anti-CD11b Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: ICRF44]
Alexa Fluor® 594 anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: IF, FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 2.5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Common Marmoset;Cynomolgus;Rhesus;Swine (Pig;Porcine)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty