Anti-CD11a Mouse Monoclonal Antibody [clone: HI111]

Supplier: BioLegend
301223 301223
10757-530EA 139.32 USD
CA10757-530 10757-530
Anti-CD11a Mouse Monoclonal Antibody [clone: HI111]
Antibodies
Purified anti-human CD11a (MaxPar® Ready) [HI111]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Capuchin monkey, Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Sheep (Ovine); Apps: FC, CyTOF®; Size: 100 μg

This product is suitable for use with the <a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the <a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: HI111
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cattle (Bovine;Cow);Capuchin monkey;Chimpanzee;Cynomolgus;Dog (Canine);Horse (Equine);Rabbit (Lapine);Sheep (Ovine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR