Anti-CD11a Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: HI111]

Supplier: BioLegend
301222 301222
10757-528EA 285.98 USD
CA10757-528 10757-528
Anti-CD11a Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: HI111]
Antibodies
Alexa Fluor® 594 anti-human CD11a [HI111]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Capuchin monkey, Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Sheep (Ovine); Apps: IF, FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 2.5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: HI111
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cattle (Bovine;Cow);Capuchin monkey;Chimpanzee;Cynomolgus;Dog (Canine);Horse (Equine);Rabbit (Lapine);Sheep (Ovine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR