Would you like to log in again?
Odyssey® TBS Blocking Buffers, LI-COR
Supplier: Li-Cor
927-50150 927-50000 927-50125 927-50100 927-50010 927-50003
102971-252 102971-244 102971-250 102971-248 102971-246 103011-502
Odyssey® TBS Blocking Buffers, LI-COR
Determining the best blocking agent for each target will provide the best chance for success
  • Contains no mammalian proteins, for the lowest cross-reactivity with mammalian antibodies
  • Ready-to-use formulation in TBS
  • Suitable for use with the widest range of targets for Western blot detection
  • Also available in kits with Immobilon®-FL PVDF membranes
  • Ideal for primary antibodies that are optimized for TBS-based buffer systems
  • Ideal for detection of phosphorylated targets
  • Excellent choice for biotin-streptavidin detection
  • Superior in performance for alkaline phosphatase detection
Some blocking buffers may interfere with detection. Milk protein blockers, for instance, may contain endogenous biotin or phosphorylated proteins, which result in high background.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty